Carbon disulfide ≥99.9%, OmniSolv®, Supelco®
Supplier: MilliporeSigma
|
Danger
|
CX0396-6
CX0396-6
CACX0396-6CS
1070
CAD
CACX0396-6
CATXCX0396-6
Carbon disulfide ≥99.9%, OmniSolv®, Supelco®
Carbon disulphide
Meets Reagent Specifications for testing USP/NF monographs
Formula:
CS₂ Boiling Pt: 46.2 °C (1013 hPa) Melting Pt: –111.6 °C Density: 1.2632 g/cm³ (20 °C) Flash Pt: –30 °C Storage Temperature: Ambient |
MDL Number:
MFCD00011321 CAS Number: 75-15-0 EINECS: 200-843-6 UN: 1131 ADR: 3,I Merck Index: 13,01821 |
|
Specification Test Results
Assay (GC) | 99.9% min |
Color (APHA) | 10 max |
Filtered through 0.2 µm filter | To pass test |
Form | Clear liquid |
Infrared spectrum | Conforms to standard |
Residue after evaporation | 5ppm max |
UV Abs. at 382 nm | 1.00AU max |
UV Abs. at 388 nm | 0.50AU max |
UV Abs. at 403 nm | 0.10AU max |
UV Abs. at 410 nm | 0.05AU max |
UV Abs. at 500 nm | 0.005AU max |
UV Cut-off | 382nm max |
Water (H2O) | 0.03% max |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...