Carbon disulfide ≥99.9%, GR ACS, Supelco®

Supplier: MilliporeSigma
Danger

CX0395-5 CX0395-6
CACX0395-5CS 4716 CAD
CACX0395-5 CACX0395-6
Carbon disulfide ≥99.9%, GR ACS, Supelco®
Carbon disulphide

Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs

Formula: CS₂
Boiling Pt: 46.2 °C (1013 hPa)
Melting Pt: –111.6 °C
Density: 1.2632 g/cm³ (20 °C)
Flash Pt: –30 °C
Storage Temperature: Ambient
MDL Number: MFCD00011321
CAS Number: 75-15-0
EINECS: 200-843-6
UN: 1131
ADR: 3,I
Merck Index: 13,01821
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a7fe0c906449b3b5461b1bac971c583c92c0b41c&vid=59e9028fbb4d1c2acb6f98101726e8ec520e0aa8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (GC) 99.9% min
Color (APHA) 10 max
Form Clear liquid
Hydrogen sulfide (H2S) To pass test (about 1.5 ppm)
Residue after evaporation 0.002% max
Sulfur dioxide (SO2) To pass test (about 2.5 ppm)
Water (H2O) 0.05% max


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR