CP100™ BT Sterile Latex Hand-Specific Gloves, Cardinal
Supplier: Cardinal
CP100™
2Y5003 2Y5000 2Y5005 2Y5004 2Y5001 2Y5007 2Y5002 2Y5000 2Y5006
CAPH2Y5000CS 619.14 CAD
PH2Y5005 CAPH2Y5000 PH2Y5004 PH2Y5003 PH2Y5002 PH2Y5001 PH2Y5000 PH2Y5007 PH2Y5006
CP100™ BT Sterile Latex Hand-Specific Gloves, Cardinal
Gloves
Sterile, clean-processed gloves feature a comfortable flex-and-stretch design and low particulate levels for minimized contamination of controlled environments.
  • Ambidextrous, beaded cuff
  • Fully textured palm and fingers provide a secure grip
  • Clean processed, powder free
The antislip, textured finish enhances the grip for better handling. Tapered fingers help reduce fatigue while providing increased tactile sensitivity. Beaded cuff with reinforced ribs prevents constriction and reduces rolldown. Hand-specific. Powder-free. Color: natural white.
Certifications: Sterilized by gamma irradiation to a Sterility Assurance Level of 10⁻⁶ in accordance with AAMI guidelines. Certificates of sterility available upon request. Each lot is performance tested and is fully traceable.
Ordering information: 50 individually packed pairs per poly bag, four poly bags per case.