Capping B 16% n-Methylimidazole in THF, Novabiochem® for DNA/RNA synthesis
Supplier: MilliporeSigma
BI0345-0450 BI0345-4000
CABI0345-0450CS 1530 CAD
CABI0345-0450 CABI0345-4000 EMBI0345-0201
Capping B 16% n-Methylimidazole in THF, Novabiochem® for DNA/RNA synthesis
Capping Reagent
These cappping reagents are involved in the formulations for oligonucleotide synthesis.
  • Soluble in water
  • Clear liquid form
  • Flammable, corrosive, and toxic effects
These are suitable for all kind of biosynthesis processes such as DNA, RNA and peptide synthesis.