Coomassie® brilliant blue R-250, OmniPur®

Supplier: MilliporeSigma

Synonyms: Serva Blue R, Brillant blue R, N-[4-[[4-[(4-Ethoxyphenyl)amino]phenyl][4-[ethyl[(3-sulfophenyl)methyl]amino]phenyl]methylene]-2,5-cyclohexadien-1-ylidene]-N-ethyl-3-sulfo-benzenemethanaminium inner salt, sodium salt (1:1), Hydrogen [4-[4-(p-ethoxyanilino)-4'-[ethyl(m-sulfonatobenzyl)amino]benzhydrylene]cyclohexa-2,5-dien-1-ylidene](ethyl)(m-sulfonatobenzyl)ammonium monosodium salt, N-[4-[[4-[(4-Ethoxyphenyl)amino]phenyl][4-[ethyl[(3-sulphophenyl)methyl]amino]phenyl]methylene]-2,5-cyclohexadien-1-ylidene]-N-ethyl-3-sulpho-benzenemethanaminium inner salt, sodium salt (1:1), Coomassie Blue R 250, Brilliant Cyanine 6B, Hydrogen [4-[4-(p-ethoxyanilino)-4'-[ethyl(m-sulphonatobenzyl)amino]benzhydrylene]cyclohexa-2,5-dien-1-ylidene](ethyl)(m-sulphonatobenzyl)ammonium monosodium salt, Acid Blue 83, Brilliant Blue R250, Brilliant Blue R, Brilliant indocyanine 6B

3340-25GM
CA-EM3340EA 131.9 CAD
CA-EM3340 CAEM3345
Coomassie® brilliant blue R-250, OmniPur®
Coomassie® Brilliant Blue R-250

Powder. Use for post-electrophoresis staining of proteins in applications requiring a high degree of sensitivity and/or solubility.

Formula: C₄₅H₄₄N₃NaO₇S₂
Melting Pt: 174…180 °C
Storage Temperature: Ambient
MDL Number: MFCD00041762
CAS Number: 6104-59-2
EINECS: 228-060-5

Order Now

Specification Test Results

Appearance Blue to red crystalline powder
Molar absorptivity Em (550 nm, water, dry basis) 33000 min


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR