Citric acid ≥99.5%, OmniPur® for biotechnology, Millipore®
Supplier: MilliporeSigma
|
Warning
|
3200-1KG
3220-5KG
CA-EM3200EA
178.23
CAD
CA-EM3200
CA-EM3220
Citric acid ≥99.5%, OmniPur® for biotechnology, Millipore®
Citric acid
For molecular biology. DNase-, RNase-, and protease-free.
Formula:
HOC(COOH)(CH₂COOH)₂ Boiling Pt: 310 °C (1013 hPa) Melting Pt: 153…159 °C (lit.) Density: 1.665 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011669 CAS Number: 77-92-9 EINECS: 201-069-1 REACH: 01-2119457026-42-0000 Merck Index: 13,02350 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...