3-[(3-Cholamidopropyl)dimethylammonio]-1-propane sulfate (CHAPS), OmniPur®, Millipore®

Supplier: MilliporeSigma

3055-100GM 3050-10GM
CA3055EA 4530.45 CAD
CA3055 CAEM3050
3-[(3-Cholamidopropyl)dimethylammonio]-1-propane sulfate (CHAPS), OmniPur®, Millipore®
3-[(3-Cholamidopropyl)dimethylammonio]-1-propane sulphate (CHAPS)

Zwitterionic detergent ideal for solubilization of membrane proteins, analytical, diagnostic, chromatography, electrophoresis work, and enzymology and immunology applications.
Suitable for Molecular Biology

Formula: C₃₂H₅₈N₂O₇S
Melting Pt: 116 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00081083
CAS Number: 75621-03-3
Merck Index: 13,02052

Order Now

Specification Test Results

Conductivity (10%, water) ≤50 µmho
DNase None detected
Protease None detected
Purity (TLC) Single spot
RNase None detected
Solubility 20% in water, passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR