CR100® Sterile Latex Gloves, Cardinal
Supplier: Cardinal Health
2Y1624 2Y1626 2Y1627 2Y1628 2Y1625 2Y1630 2Y1640
CAPH2Y1624CS 372.3 CAD
CAPH2Y1624 CAPH2Y1626 CAPH2Y1627 CAPH2Y1628 CAPH2Y1625 CAPH2Y1630 CAPH2Y1640
CR100® Sterile Latex Gloves, Cardinal
Gloves
Versatile, high-quality latex gloves can be used in controlled environments.
  • Sterile, clean processed, powder free
  • Comfortable flex-and-stretch design
  • Anti-slip, textured finish enhances the grip for better handling
  • Beaded cuff with reinforced ribs prevents constriction and reduces rolldown
Hand-specific and powder-free. Bisque finish, tapered cuff. Sterilized by gamma radiation to a Sterility Assurance Level of 10⁻⁶ in accordance with AAMI guidelines.
Ordering information: Pair packed and double-bagged for superior protection.