Sterile Latex Gloves, Kimtech™ G3, Kimberly-Clark
Supplier: Microflex
56849 56842 56848 56847 56846 56845 56844 56843
CA32933-743CS 667.43 CAD
CA32933-743 CA32933-745 CA32933-879 CA32933-881 CA32933-883 CA32933-885 CA32933-887 CA32933-889
Sterile Latex Gloves, Kimtech™ G3, Kimberly-Clark
Gloves
Ideal for critical environments. Hand-specific gloves are specially washed to control particle levels.
  • Low protein and endotoxin
  • Sterilized by gamma radiation
  • Traceability system and irradiation certifications meet internal validation requirements
Textured, non-slip surface provides exceptional wet/dry grip without compromising tactile sensitivity.
The tapered, beaded cuff reduces rolldown, makes donning easier, enhances cuff strength, and forms a better seal on the garment.
Recommended for ISO Class 4 (FED-STD-209E Class 1/M1.5) or higherMore Product Information