Cyclopentane ≥99.9%, B&J Brand™ for HPLC, for gas chromatography, for spectrophotometry, Burdick & Jackson™
Supplier: HONEYWELL BURDICK & JACKSON CA
|
Danger
|
Synonyms:
Pentamethylene
057-4
057-4
CABJ057-4CS
2648.62
CAD
CABJ057-4
BJ057-4
Cyclopentane ≥99.9%, B&J Brand™ for HPLC, for gas chromatography, for spectrophotometry, Burdick & Jackson™
Cyclopentane
For HPLC, liquid and gas chromatography, pesticide residue analysis, spectophotometry, organic synthesis and combinatorial chemistry.
- Minimal UV contaminants
- High GC purity
- Low water content
- Outstanding lot-to-lot consistency
Formula:
C₅H₁₀ MW: 70.13 g/mol Boiling Pt: 49.3 °C Melting Pt: –93.9 °C Density: 0.745 g/cm³ (20 °C) Flash Pt: –7 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001356 CAS Number: 287-92-3 UN: 1146 ADR: 3,II Merck Index: 13,02769 |
Specification Test Results
Electron Capture GC | 10 ng/L max. |
Water by Karl Fischer Titration | 0.01 % max. |
GC Analysis (Cyclopentane) | 65 % min. |
GC Analysis (total C5 and C6 isomers) | 99.9 % min. |
Refractive Index (20ºC) | 1.3930 - 1.3970 |
Residue | 1 mg/L max. |
UV Absorbance @ 215 nm | 0.5 AU max. |
UV Absorbance @ 225 nm | 0.1 AU max. |
UV Absorbance @ 300 nm | 0.005 AU max. |
UV Cutoff | 198 nm max. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...