Cyclohexane 99,5, B&J Brand™ ACS HPLC, Spectrophotometry, Burdick & Jackson™
Supplier: HONEYWELL BURDICK & JACKSON CA
|
Danger
|
Synonyms:
Hexahydrobenzene, Hexamethylene
053-1L
053-4
CABJ053-1LCS
778.69
CAD
CABJ053-1L
CABJ053-4
Cyclohexane 99,5, B&J Brand™ ACS HPLC, Spectrophotometry, Burdick & Jackson™
Cyclohexane
For HPLC, liquid and gas chromatography, pesticide residue analysis, spectophotometry, organic synthesis and combinatorial chemistry.
- Minimal UV contaminants
- High GC purity
- Low water content
- Outstanding lot-to-lot consistency
Formula:
C₆H₁₂ MW: 84.16 g/mol Boiling Pt: 81 Melting Pt: 7 Density: 0.78 g/cm³ (20 °C) Flash Pt: –20 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003814 CAS Number: 110-82-7 EINECS: 203-806-2 UN: 1145 ADR: 3 II Merck Index: 13,02752 |
|
Specification Test Results
Electron Capture GC | 10 ng/L max. |
Water by Karl Fischer | 0.01 % max. |
GC Analysis | 99.5 % min. |
Refractive Index (20ºC) | 1.4239 - 1.4279 |
Residue | 1 mg/L max. |
UV Absorbance @ 225 nm | 0.17 AU max. |
UV Absorbance @ 250 nm | 0.02 AU max. |
UV Absorbance @ 300 nm | 0.005 AU max. |
UV Absorbance @ 400 nm | 0.005 AU max. |
UV Cutoff | 200 nm max. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...