CE5 System Latex Gloves, Microflex®, Ansell
Supplier: Microflex
CE5-512-S CE5-512-M CE5-512-XL
CA32916-710CS 2701.6 CAD
CA32916-710 CA32916-712 CA32916-714 CA32916-716
CE5 System Latex Gloves, Microflex®, Ansell
Gloves
These ambidextrous gloves are for use in ISO Class 5 (FED-STD-209E Class 100/M3.5) controlled environments.
  • Low surface particulates, ionic extractables, and non-volatile residues
  • Silicone-free
  • Micro-textured surface enhances wet or dry grip
  • Provides superb aesthetics, dexterity, tactile sense, fit, feel, and comfort
  • Powder-free
Textured, ambidextrous gloves are compatible for use with both wet and dry environments.
Certifications: Lot-specific Certificate of Analysis available upon request.
Packaging: Packed 100 gloves per vacuum-sealed double poly bag, 10 poly bags per case.
Caution: These gloves contain natural rubber latex which may cause allergic reactions. Safe use of this glove by or on latex-sensitized individuals has not been established.