Chloroform ≥99.8% stabilized ACS, VWR Chemicals BDH®

Supplier: VWR International
Danger


Stabilization: Stabilised with amylene

BDH1109-204L BDH1109-4LG BDH1109-19L
BDH1109-204LEA 11022.27 CAD
BDH1109-204L BDH1109-4LG BDH1109-19L
Chloroform ≥99.8% stabilized ACS, VWR Chemicals BDH®
Chloroform

Stabilized with amylene.

Formula: CHCl₃
Boiling Pt: 61.7 °C (1013 hPa)
Melting Pt: –63 °C
Density: 1.4832 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000826
CAS Number: 67-66-3
EINECS: 200-663-8
UN: 1888
ADR: 6.1,III
REACH: 05-2114614906-45
Merck Index: 13,02160
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9cc00da8cefd4e04c0ea1d7c5801308cd73eeef9&vid=c7d46a2ee73651eae0f3654c272e820106b2f04f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear, colorless liquid
Water (K.F.) Less than 0.020%
Residue Less than 0.001% (0.0010g/67 mL)
GC Purity Greater than 99.8%
Amylene (Capillary GC-FID) Record value
Color (APHA) Less than 10
Acetone and aldehyde Passes test (less than 0.005% as acetone. No turbidity should develop.)
Acid and chloride Passes test (Blue litmus paper should not change color. No turbidity should be produced.)
Free chlorine (Cl) Passes test (Lower layer should not show a violet tint.)
Lead (Pb) Less than 0.05 ppm (Purplish hue of sample should not exceed that of the standard.)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR