Cyclopentylmethanol 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Cyclopentanemethanol

A11595-06 A11595-14 A11595-22
CAAAA11595-06EA 127.03 CAD
CAAAA11595-06 CAAAA11595-14 CAAAA11595-22
Cyclopentylmethanol 98%
Cyclopentylmethanol

Purity: 98% CAS No.: 3637-61-4 Molecular Formula: C6H12O Formula Weight: 100.16 Flash Point: 62°(143°F) Refractive Index: 1.4580 Boiling Point: 162-163° Beilstein Reference: 1919000 MDL No.: MFCD00001384

Formula: C₆H₁₂O
MW: 100.16 g/mol
Boiling Pt: 162…163 °C
Density: 0.925
Flash Pt: 62 °C(143 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001384
CAS Number: 3637-61-4
EINECS: 222-861-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR