Copper ≥99.9% (metals basis), wire, Length: 1.5mm (0.06in), Ø 1.5 mm (0.06 in)

Supplier: Thermo Scientific Chemicals

44549-G9 44549-H4
CAAA44549-G9EA 178.54 CAD
CAAA44549-G9 CAAA44549-H4 AA44549-G5
Copper ≥99.9% (metals basis), wire, Length: 1.5mm (0.06in), Ø 1.5 mm (0.06 in)
Copper

MDL: MFCD00010965


Purity: 99.9% CAS No.: 7440-50-8 Physical Form: ≈15.8g/m Odor: Odorless MDL No.: MFCD00010965

Formula: Cu
MW: 63.55 g/mol
Boiling Pt: 2595
Melting Pt: 1083
Density: 8.94 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010965
CAS Number: 7440-50-8
EINECS: 231-159-6
Merck Index: 13,02545

Order Now

Specification Test Results

Assay (by Metals Basis) 99.9%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR