Cobalt ≥99.998% (metals basis), powder, Puratronic® -22 mesh

Supplier: Thermo Scientific Chemicals
Danger

10610-06 10610-14
CAAA10610-06EA 172.22 CAD
CAAA10610-06 CAAA10610-14
Cobalt ≥99.998% (metals basis), powder, Puratronic® -22 mesh
Cobalt

MDL: MFCD00010935


Purity: 99.998% CAS No.: 7440-48-4 Appearance: Black Odor: Odorless UN No.: 3089 MDL No.: MFCD00010935

Formula: Co
MW: 58.93 g/mol
Boiling Pt: 2870
Melting Pt: 1495
Storage Temperature: Ambient
MDL Number: MFCD00010935
CAS Number: 7440-48-4
EINECS: 231-158-0
UN: 3089
ADR: 4.1 III
Merck Index: 12,02488

Order Now

Specification Test Results

Assay (by Metals Basis) 99.998%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR