PFE Latex Examination Gloves, Kimberly-Clark
Supplier: Microflex
57110 57220 57550 57330
CA32934-003CS 291.4 CAD
CA32934-003 CA32934-007 CA32934-011 CA32934-015 CA32934-019 32934-007 32934-011 32934-015
PFE Latex Examination Gloves, Kimberly-Clark
Gloves
Gloves provide protection from infectious fluids and bloodborne pathogens.
  • Ambidextrous
  • Beaded cuff
  • For single use only
  • AQL 1.5
  • Thickness: 6.7 mil (finger)
Fully textured finish provides excellent grip in both wet and dry situations. Gloves are powder-free to minimize powder-associated transfer of latex proteins and chemical allergens.
Caution: This product contains natural rubber latex, which may cause allergic reactions. Safe use of this glove by or on latex-sensitive individuals has not been established. This product should not be worn by orMore Product Information
Order Now
SPECIFICATIONS
Ambidextrous/Hand-specific Ambidextrous Ambidextrous Ambidextrous
Environmentally Preferable
Disposable / reusable Disposable Disposable Disposable
Latex/Latex-free Latex Latex Latex
Powdered/Powder-free Powder-free Powder-free Powder-free
Cuff thickness 4.3 mil 4.3 mil 4.3 mil
Finger thickness 6.7 mil 6.7 mil 6.7 mil
Palm thickness 5.5 mil 5.5 mil 5.5 mil