m-Xylene-4-sulfonic acid hydrate ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
D0680-25G
D0680-500G
TCD0680-025GEA
94.8
CAD
TCD0680-025G
TCD0680-500G
m-Xylene-4-sulfonic acid hydrate ≥98.0% (by HPLC, titration analysis)
m-Xylene-4-sulfonic acid hydrate
CAS Number: 88-61-9
MDL Number: MFCD00007483
Molecular Formula: C8H10O3S
Molecular Weight: 186.23
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 61
Other Product Names / Synonyms: 2,4-Dimethylbenzenesulfonic Acid
Formula:
C₈H₁₀O₃S Melting Pt: 61 °C |
MDL Number:
MFCD00007483 CAS Number: 88-61-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...