o-Xylene ≥98.0% (by GC), Supelco®

Supplier: MilliporeSigma
Danger

XX0020-5 XX0020-1
CAXX0020-5EA 275 CAD
CAXX0020-5 CAXX0020-1
o-Xylene ≥98.0% (by GC), Supelco®
o-Xylene

Reagent.

Formula: C₈H₁₀
Boiling Pt: 144 °C (1013 hPa)
Melting Pt: –25 °C
Density: 0.88 g/cm³ (20 °C)
Flash Pt: 30 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00008519
CAS Number: 95-47-6
EINECS: 202-422-2
UN: 1307
ADR: 3,III
Merck Index: 12,10214
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5dd6c62d1c62b6e7f71df7e157aae77162b3bcf1&vid=3ec8c09bb94f634e9f90b6f9e8320cd626f276d0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (GC) 98.0% min
Color (APHA) 10 max
Form Clear liquid
Infrared spectrum Conforms to standard


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR