m-Xylene 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 1,3-Dimethylbenzene

L03788-0F L03788-AE L03788-AP
CAAAAL03788-0FEA 281.24 CAD
CAAAAL03788-0F CAAAAL03788-AE CAAAAL03788-AP
m-Xylene 99%
m-Xylene

Liquid. Miscible with many organic solvents, including alcohol and ether.


Purity: 99% CAS No.: 108-38-3 Molecular Formula: C8H10 Formula Weight: 106.17 Flash Point: 25°(77°F) Refractive Index: 1.4970 Odor: Mild Melting Point: -47° Boiling Point: 138-139° Beilstein Reference: 605441 Merck Reference: 14,10081 UN No.: 1307 MDL No.: MFCD00008536

Formula: C₈H₁₀
Boiling Pt: 138…139 °C
Melting Pt: –47 °C
Density: 0.866
Flash Pt: 25 °C (77 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008536
CAS Number: 108-38-3
EINECS: 203-576-3
UN: 1307
ADR: 3,III
Merck Index: 14,10081

Order Now

Specification Test Results

Boiling Point 138–139°C (280–282°F)
Flash Point 25°C (77°F)
Melting Point –47°C (–53°F)
Refractive Index 1.497


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR