Xenon difluoride ≥99.5% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Danger
|
39739-04
39739-09
39739-18
CAAA39739-04EA
281.24
CAD
CAAA39739-04
CAAA39739-09
CAAA39739-18
Xenon difluoride ≥99.5% (metals basis)
Xenon difluoride
Crystalline granules. Moisture sensitive. Fluorinating agent. Purity based on metal contaminates only. Shipped in fluoropolymer resin.
Purity: 99.5%
CAS No.: 13709-36-9
Molecular Formula: XeF2
Formula Weight: 169.30
Physical Form: Crystalline Granules, shipped in teflon
Density: 4.32
Appearance: White
Melting Point: 127-129°
Merck Reference: 14,10072
UN No.: 3087
MDL No.: MFCD00040538
Fluorinating agent
Formula:
F₂Xe Boiling Pt: 114 °C (1013 hPa) Melting Pt: 127…129 °C Density: 4.32 Storage Temperature: Ambient |
MDL Number:
MFCD00040538 CAS Number: 13709-36-9 EINECS: 237-251-2 UN: 3087 ADR: 5.1,II Merck Index: 14,10072 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...