Xylene (mixture of isomers), Epredia™ ACS, Reagent Grade

Supplier: RICHARD-ALLAN SCIENTIFIC CA
Danger

Synonyms: Dimethylbenzene

9900-1 9900-5
CA48218-746CS 540.58 CAD
CA48218-746 CA48218-748
Xylene (mixture of isomers), Epredia™ ACS, Reagent Grade
Xylene (mixture of isomers)

Ensure high-quality and consistent results with reagent grade Epredia™ Xylenes (ACS).

Formula: C₆H₄(CH₃)₂
MW: 106.17 g/mol
Boiling Pt: 140
Melting Pt: –34 °C
Density: 0.86…0.88 g/cm³ (20 °C)
Flash Pt: 25
Storage Temperature: Ambient
MDL Number: MFCD00077264
CAS Number: 1330-20-7
EINECS: 215-535-7
UN: 1307
ADR: 3 III
Merck Index: 13,10136
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2deefc7b7b67c10e64003e243ff3216fad27cf7d&vid=9fa01ddafd4814d5f34266ae7933463a29d92d19&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR