2,2-Bis(3-amino-4-hydroxyphenyl)hexafluoropropane ≥98.0%

Supplier: TCI America

Synonyms: 4,4'-(Hexafluoroisopropylidene)bis(2-aminophenol), 4,4'-(Perfluoropropane-2,2-diyl)bis(2-aminophenol)

B1415-5G B1415-25G
TCB1415-005GEA 137.46 CAD
TCB1415-005G TCB1415-025G
2,2-Bis(3-amino-4-hydroxyphenyl)hexafluoropropane ≥98.0%
2,2-Bis(3-amino-4-hydroxyphenyl)hexafluoropropane

CAS Number: 83558-87-6
MDL Number: MFCD00039147
Molecular Formula: C15H12F6N2O2
Molecular Weight: 366.26
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 243


Product Categories: Materials Chemistry, Monomers, Monomers, Polyimide Raw Materials, Bisphenol AF type Compounds (for High-Performance Polymer Research)
Other Product Names / Synonyms: 4,4'-(Hexafluoroisopropylidene)bis(2-aminophenol)

Formula: C₁₅H₁₂F₆N₂O₂
MW: 366.26 g/mol
Melting Pt: 243 °C
MDL Number: MFCD00039147
CAS Number: 83558-87-6

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR