Behenic acid ≥95.0%

Supplier: TCI America

Synonyms: Docosanoic acid

B1248-5G B1248-25G
TCB1248-5GEA 49.61 CAD
TCB1248-5G TCB1248-025G
Behenic acid ≥95.0%
Behenic acid

CAS Number: 112-85-6
MDL Number: MFCD00002807
Molecular Formula: C22H44O2
Molecular Weight: 340.59
Purity/Analysis Method: >95.0% (GC)
Form: Crystal
Boiling point (°C): 306
Melting point (°C): 80
Flash Point (°C): 228


Product Categories: Materials Chemistry, Pressure & Heat Sensitive Recording Materials, Bioscience, Lipids, Higher Fatty Acids & Higher Alcohols
Other Product Names / Synonyms: Docosanoic Acid

Formula: C₂₂H₄₄O₂
MW: 340.59 g/mol
Boiling Pt: 306
Melting Pt: 80
Density: 0.8221 g/cm³ (20 °C)
Flash Pt: 228 °C
Storage Temperature: Ambient
MDL Number: MFCD00002807
CAS Number: 112-85-6
EINECS: 204-010-8
Merck Index: 13,01024
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a8189eef0d684b7933dd96f3144cf210e4923bd8&vid=00f96c11ebe178bc43709daf5174d2e6ad682ae1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR