3-Methyl-1-phenyl-2-butanone ≥98.0%

Supplier: TCI America

B1029-25ML B1029-5ML
TCB1029-25MLEA 370.19 CAD
TCB1029-25ML TCB1029-005ML
3-Methyl-1-phenyl-2-butanone ≥98.0%
3-Methyl-1-phenyl-2-butanone

CAS Number: 2893-05-2
MDL Number: MFCD00040487
Molecular Formula: C11H14O
Molecular Weight: 162.23
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 240
Specific Gravity (20/20): 0.97


Other Product Names / Synonyms: Benzyl Isopropyl Ketone

Formula: (C₆H₅)CH₂C(O)CH(CH₃)CH₃
Boiling Pt: 240 °C
Density: 0.97 (20 °C)
MDL Number: MFCD00040487
CAS Number: 2893-05-2
EINECS: 220-765-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0b23957620c924480c13566dca7c5fa590895368&vid=13d1c7db71fe291fb86c04321823fa518510eafb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR