5-tert-Butyl-m-xylene ≥98.0%

Supplier: TCI America

B0965-25ML B0965-500ML
TCB0965-025MLEA 36.34 CAD
TCB0965-025ML TCB0965-500ML
5-tert-Butyl-m-xylene ≥98.0%
5-tert-Butyl-m-xylene

CAS Number: 98-19-1
MDL Number: MFCD00008832
Molecular Formula: C12H18
Molecular Weight: 162.28
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 202
Flash Point (°C): 72
Specific Gravity (20/20): 0.87


Other Product Names / Synonyms: 1-tert-Butyl-3,5-dimethylbenzene

Formula: C₁₂H₁₈
Boiling Pt: 202 °C
Melting Pt: –22 °C
Density: 0.87 (20 °C)
Flash Pt: 72 °C
Storage Temperature: Ambient
MDL Number: MFCD00008832
CAS Number: 98-19-1
EINECS: 202-647-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3dbad6acf57915c2a7e7c2d0a991f7ddcc237b9e&vid=3b808698fbafe13799066bda835d9fa5f9562fd6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR