N-Butyl-N-(4-hydroxybutyl)nitrosamine ≥90.0%
Supplier: TCI America
Synonyms:
1-Butanol, 4-(butylnitrosoamino)-, N-Butyl-N-nitroso-4-aminobutanol, N-Butyl-N-butan-4-ol-nitrosamine, BBN
CAS Number: 3817-11-6
MDL Number: MFCD00059008
Molecular Formula: C8H18N2O2
Molecular Weight: 174.24
Purity/Analysis Method: >90.0% (GC)
Form: Clear Liquid
Boiling point (°C): 167
Specific Gravity (20/20): 1.01
Storage Temperature: 0-10°C
Product Categories: Bioscience, Reagents for Carcinogenic Models
Other Product Names / Synonyms: N-Butyl-N-butan-4-ol-nitrosamine, N-Butyl-N-nitroso-4-aminobutanol, BBN
Formula:
C₈H₁₈N₂O₂ Boiling Pt: 167 °C Density: 1.01 (20 °C) |
MDL Number:
MFCD00059008 CAS Number: 3817-11-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...