1-Bromopentadecane ≥98.0%

Supplier: TCI America

B0936-25ML B0936-250ML
TCB0936-025MLEA 178.43 CAD
TCB0936-025ML TCB0936-250ML
1-Bromopentadecane ≥98.0%
1-Bromopentadecane

CAS Number: 629-72-1
MDL Number: MFCD00000229
Molecular Formula: C15H31Br
Molecular Weight: 291.32
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 158
Flash Point (°C): 110
Freezing point (°C): 18
Specific Gravity (20/20): 1.01


Product Categories: Materials Chemistry, Derivatizing Reagents for Solubility Improvement
Other Product Names / Synonyms: Pentadecyl Bromide

Formula: BrH₂C(CH₂)₁₃CH₃
Boiling Pt: 158 °C
Melting Pt: 19 °C
Density: 1.01 (20 °C)
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00000229
CAS Number: 629-72-1
EINECS: 211-104-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d77fcb8a09f53c4cd92d957a17e1047026ecfc04&vid=e8a0959f07495bbb5bdfbe0a59a27aa8b0ad2d2d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR