Benzoin methyl ether ≥98.0%

Supplier: TCI America

Synonyms: 1,2-Diphenyl-2-methoxyethan-1-one, 2-Methoxy-1,2-diphenylethanone

B0869-500G B0869-25G
TCB0869-500GEA 900.6 CAD
TCB0869-500G TCB0869-025G
Benzoin methyl ether ≥98.0%
Benzoin methyl ether

CAS Number: 3524-62-7
MDL Number: MFCD00008492
Molecular Formula: C15H14O2
Molecular Weight: 226.28
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 189
Melting point (°C): 50
Flash Point (°C): 110


Product Categories: Materials Chemistry, Polymerization Initiators
Other Product Names / Synonyms: 2-Methoxy-2-phenylacetophenone

Formula: C₁₅H₁₄O₂
MW: 226.27 g/mol
Boiling Pt: 189 °C
Melting Pt: 50 °C
Flash Pt: 110 °C
MDL Number: MFCD00008492
CAS Number: 3524-62-7
EINECS: 222-538-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d4f8bfb7b35faa3b64d8d71b05258f7e603b05db&vid=d4f8bfb7b35faa3b64d8d71b05258f7e603b05db&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR