4-Bromopyridinium chloride ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Warning

B0653-5G B0653-25G
TCB0653-005GEA 126.4 CAD
TCB0653-005G TCB0653-025G
4-Bromopyridinium chloride ≥98.0% (by titrimetric analysis)
4-Bromopyridinium chloride

CAS Number: 19524-06-2
MDL Number: MFCD00012828
Molecular Formula: C5H4BrN
Molecular Weight: 194.46
Purity/Analysis Method: >98.0% (T)
Form: Crystal

Formula: C₅H₅BrClN
Melting Pt: 235…238 °C
MDL Number: MFCD00012828
CAS Number: 19524-06-2
EINECS: 243-128-4
UN: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=41a6102314bfa63afac590d5be7fa8a271cd8968&vid=41a6102314bfa63afac590d5be7fa8a271cd8968&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR