Orthoboric acid 4% (w/v) aqueous solution with bromocresol green/ methyl red indicator for Kjeldahl nitrogen analysis

Supplier: Ricca Chemical
Danger

Synonyms: Boron trihydroxide, Boron hydroxide, Boric acid, Boracic acid

R1070000-4A R1070000-10F R1070000-20F R1070000-1A
RC10701EA 214.88 CAD
RC10701 RC107025 RC10705 RCR1070000-1A
Orthoboric acid 4% (w/v) aqueous solution with bromocresol green/ methyl red indicator for Kjeldahl nitrogen analysis
ortho-Boric acid

With bromocresol green-methyl red indicator.

Formula: B(OH)₃
MW: 61.83 g/mol
Boiling Pt: 185
Melting Pt: 160
Storage Temperature: Ambient
MDL Number: MFCD00011337
CAS Number: 10043-35-3
EINECS: 233-139-2
Merck Index: 13,01326
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=098dd700154daaf6338ebb270902bb2bc704f27d&vid=6eec7d529090dd4c4b966ed8cd8d057153c5a47b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Red-orange liquid
Suitability for Use Green (Basic) - Grey (Neutral) - Red (Acidic)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR