Bis(4-hydroxyphenyl) sulfone ≥98.0%

Supplier: TCI America

Synonyms: 4-Hydroxyphenyl sulfone, 4,4'-Sulphonyldiphenol, 4,4'-Dihydroxy diphenyl sulphone, 4,4'-Dihydroxy diphenyl sulfone, 4,4'-sulfonyldiphenol, 4,4'-Sulfonyldiphenol, 4-Hydroxyphenyl sulphone, 4,4'-sulphonyldiphenol

B0495-25G B0495-500G
TCB0495-025GEA 33.73 CAD
TCB0495-025G TCB0495-500G
Bis(4-hydroxyphenyl) sulfone ≥98.0%
Bis(4-hydroxyphenyl) sulphone

CAS Number: 80-09-1
MDL Number: MFCD00002350
Molecular Formula: C12H10O4S
Molecular Weight: 250.27
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 247
Lambda max.: 295 nm (H2O)


Product Categories: Materials Chemistry, Monomers, Diphenyl Sulfones (for High-Performance Polymer Research), Pressure & Heat Sensitive Recording Materials
Other Product Names / Synonyms: Bisphenol S, 4,4'-Dihydroxydiphenyl Sulfone, 4,4'-Sulfonyldiphenol

Formula: C₁₂H₁₀O₄S
Melting Pt: 247 °C
MDL Number: MFCD00002350
CAS Number: 80-09-1
EINECS: 201-250-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f730bdfad5cc46d964d0b1248e999844ef9fdf62&vid=f730bdfad5cc46d964d0b1248e999844ef9fdf62&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR