4-tert-Butylcyclohexanol ≥98.0%
Supplier: TCI America
B0383-25G
B0383-500G
TCB0383-025GEA
61.07
CAD
TCB0383-025G
TCB0383-500G
4-tert-Butylcyclohexanol ≥98.0%
4-tert-Butylcyclohexanol
CAS Number: 98-52-2
MDL Number: MFCD00001473
Molecular Formula: C10H20O
Molecular Weight: 156.27
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 68
Flash Point (°C): 105
Other Product Names / Synonyms: Hexahydro-p-tert-butylphenol
Formula:
C₁₀H₂₀O Boiling Pt: 214 °C (1013 hPa) Melting Pt: 68 °C Density: 0.87 g/cm³ (80 °C) Flash Pt: 105 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001473 CAS Number: 98-52-2 EINECS: 202-676-4 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...