Orthoboric acid ≥99.5%, granular, GenAR® NF, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC

Synonyms: Boric acidBoron hydroxideBoron trihydroxideBoracic acid

Management of Change (MOC) category = R

7779-06 MACR7779-26 MACR7779-26 MACR7779-06 7779-26
CAMK777906EA 1009.75 CAD
MK7779-26 CAMK777906 JT7779-26 CAJT7779-26 MK777906
Orthoboric acid ≥99.5%, granular, GenAR® NF, Macron Fine Chemicals™
ortho-Boric acid

GMP manufactured.

Formula: B(OH)₃
MW: 61.83 g/mol
Boiling Pt: 185 °C (1013 hPa)
Melting Pt: 160 °C
Density: 1.44 g/cm³ (25 °C)
MDL Number: MFCD00011337
CAS Number: 10043-35-3
Merck Index: 13,01326
https://ca.vwr.com/stibo/web/std.lang.all/05/54/16640554.jpg

Order Now

Specification Test Results

Meets B.P. Chemical Specifications
Meets E.P. Chemical Specifications
Meets N.F. Requirements
Meets J.P. Chemical Specifications
GMP Manufactured Product
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical
NF - Assay (H₃BO₃) (dried basis) 99.5 - 100.5 %
NF - Completeness of Solution Passes Test
NF - Identification Passes Test
NF - Loss on Drying ≤0.5 %
NF - Solubility in Alcohol Passes Test
NF - LAL Bacterial Endotoxins (EU/g) ≤35
EP/BP - Assay (H₃BO₃) 99.0 - 100.5 %
EP/BP - Appearance of Solution Passes Test
EP/BP - Identification A Passes Test
EP/BP - Identification B Passes Test
EP/BP - pH of a 3.3% Solution 3.8 - 4.8
EP/BP- Organic Matter Passes Test
EP/BP - Solubility in Alcohol Passes Test
EP/BP - Sulfate (SO₄) ≤450 ppm
JP - Assay (H₃BO₃) (dried basis) 99.5 - 101.0 %
JP - Identification Passes Test
JP - Clarity and Color of Solution Passes Test
JP - Heavy Metals (as Pb) ≤10 ppm
JP - Arsenic (As) ≤5 ppm
JP - Loss on Drying ≤0.5 %
No Class 1,2,3 or other solvents are used or produced in the
manufacturing or purification of the product.
Not for Internal Use
Elemental Impurities (USP 232, EP 5.20) - Information on elemental
impurities for this product is available on the associated Product
Regulatory Data Sheet and elemental impurity profile report.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR