Orthoboric acid ≥99.5%, granular NF, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger


Management of Change (MOC) category = R

MACR1394-20 MACR1394-26
CAMK139420EA 2394.14 CAD
CAMK139420 CAMK1394-26
Orthoboric acid ≥99.5%, granular NF, Macron Fine Chemicals™
ortho-Boric acid

GMP manufactured.

Formula: B(OH)₃
Boiling Pt: 185 °C (1013 hPa)
Melting Pt: 160 °C
Density: 1.44 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011337
CAS Number: 10043-35-3
EINECS: 233-139-2
REACH: 01-2119486683-25
Merck Index: 13,01326
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=098dd700154daaf6338ebb270902bb2bc704f27d&vid=6eec7d529090dd4c4b966ed8cd8d057153c5a47b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

GMP Manufactured Product
Meets N.F. Requirements
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical
NF - Assay (H₃BO₃) 99.5 - 100.5 %
NF - Completeness of Solution Passes Test
NF - Identification Passes Test
NF - Loss on Drying ≤0.5 %
NF - Solubility in Alcohol Passes Test
Color of Solution Passes Test
No Class 1,2,3 or other solvents are used or produced in the
manufacturing or purification of the product.
Not for Internal Use
Meets Reagent Specifications for testing USP/NF monographs
Elemental Impurities (USP 232, EP 5.20) - Information on elemental
impurities for this product is available on the associated Product
Regulatory Data Sheet and elemental impurity profile report.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR