Orthoboric acid ≥99.5%, granular, AR® ACS, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

MACR2549-06 MACR2549-20 MACR2549-26
CAMK254906CS 3188.87 CAD
CAMK254906 CAMK254920 CAMK2549-26
Orthoboric acid ≥99.5%, granular, AR® ACS, Macron Fine Chemicals™
ortho-Boric acid

White granules. Toxic.

Formula: B(OH)₃
Boiling Pt: 185 °C (1013 hPa)
Melting Pt: 160 °C
Density: 1.44 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011337
CAS Number: 10043-35-3
EINECS: 233-139-2
REACH: 01-2119486683-25
Merck Index: 13,01326
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=098dd700154daaf6338ebb270902bb2bc704f27d&vid=6eec7d529090dd4c4b966ed8cd8d057153c5a47b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
ACS - Assay (H₃BO₃) ≥99.5 %
ACS - Appearance (white granules) Passes Test
ACS - Calcium (Ca) ≤0.005 %
ACS - Chloride (Cl) ≤0.001 %
ACS - Heavy Metals (as Pb) ≤0.001 %
ACS - Insoluble in Methanol ≤0.005 %
ACS - Iron (Fe) ≤0.001 %
ACS - Nonvolatile with Methanol ≤0.05 %
ACS - Phosphate (PO₄) ≤0.001 %
Solubility Passes Test
ACS - Sulfate (SO₄) ≤0.01 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR