1-Butanol ≥99.4% (by GC, corrected for water content), AR® ACS, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Warning

Synonyms: 1-Butyl alcohol, Butyl Alcohol, 1-Hydroxy butane, Propyl carbinol, n-Butanol, n-Butyl Alcohol

Management of Change (MOC) category = N

3000-04 3000-08 MACR3000-26
MK300004CS 3610.9 CAD
MK300004 MK300008 CAMK300004 CAMK300008 CAMK300026
1-Butanol ≥99.4% (by GC, corrected for water content), AR® ACS, Macron Fine Chemicals™
1-Butanol

AR—The standard Macron Fine Chemicals™ grade of analytical reagents; suitable for laboratory and general use. If the reagent also meets the requirements of the American Chemical Society Committee on Analytical Reagents, it will be denoted as an AR (ACS) r

Formula: H₃C(CH₂)₂CH₂OH
MW: 74.12 g/mol
Boiling Pt: 117.6
Melting Pt: –89 °C
Density: 0.812 g/cm³ (20 °C)
Flash Pt: 37 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00002964
CAS Number: 71-36-3
EINECS: 200-751-6
UN: 1120
ADR: 3 III
REACH: 01-2119484630-38
Merck Index: 13,01539
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e9395eeb6150a74222da52460f53cedeccd93442&vid=fd8352a556914c5be7213c576355a88e130d1a7b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
ACS - Assay (CH₃CH₂CH₂CH₂OH) (by GC, corrected for water) ≥99.4 %
ACS - Butyl Ether (C₈H₁₈O)(by GC) ≤0.2 %
ACS - Carbonyl Compounds (as Butyraldehyde) ≤0.01 %
ACS - Color (APHA) ≤10
ACS - Residue after Evaporation ≤0.005 %
ACS - Titrable Acid (as CH₃COOH) (meq/g) ≤0.0008
ACS - Water (H₂O)(by Karl Fischer titrn) ≤0.1 %
Appearance (clear, colorless liquid)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR