1-Butanol ≥99.4%, PHOTREX® for spectrophotometry, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC

Synonyms: n-ButanolPropyl carbinol1-Butyl alcoholButyl Alcohol1-Hydroxy butanen-Butyl Alcohol

Management of Change (MOC) category = N

9189-01 BAKR9189-01
CAJT9189-1CS 3009.44 CAD
JT9189-1 CAJT9189-1
1-Butanol ≥99.4%, PHOTREX® for spectrophotometry, J.T.Baker®
1-Butanol

For spectrophotometry. Lot analysis on label.

Formula: H₃C(CH₂)₂CH₂OH
MW: 74.12 g/mol
Boiling Pt: 117.6 °C (1013 hPa)
Melting Pt: –89 °C
Density: 0.812 g/cm³ (20 °C)
Flash Pt: 30 °C
MDL Number: MFCD00002964
CAS Number: 71-36-3
UN: 1120
ADR: 3,III
Merck Index: 13,01539
https://ca.vwr.com/stibo/web/std.lang.all/07/44/730744.jpg

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
ACS - Assay (CH₃CH₂CH₂CH₂OH) (by GC, corrected for water) ≥99.4 %
ACS - Color (APHA) ≤10
ACS - Residue after Evaporation ≤0.0005 %
ACS - Titrable Acid (µeq/g) ≤0.8
ACS - Butyl Ether (C₈H₁₈O)(by GC) ≤0.2 %
Carbonyl Compounds - Butyraldehyde ≤0.01 %
Water (by KF, coulometric) ≤0.05 %
Ultraviolet Absorbance (1.00-cm cell vs. water) - 400-300 nm ≤0.01
Ultraviolet Absorbance (1.00-cm cell vs. water) - 280 nm ≤0.02
Ultraviolet Absorbance (1.00-cm cell vs. water) - 260 nm ≤0.04
Ultraviolet Absorbance (1.00-cm cell vs. water) - 240 nm ≤0.10
Ultraviolet Absorbance (1.00-cm cell vs. water) - 220 nm ≤0.50
Ultraviolet Absorbance (1.00-cm cell vs. water) - 215 nm ≤1.00
Product Information (not specifications):
Boiling Point (typical)................. 117.7 °C.
Windows of Infrared Transmittance (0.1-mm path, 50-100% T), µm:
2.0 - 2.8 ; 4.3 - 6.0

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR