Benzyl alcohol ≥99.5%, BAKER ANALYZED®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Warning

Synonyms: Benzenemethanol, Phenylmethanol, Phenylcarbinol

Management of Change (MOC) category = N

9050-01 BAKR9050-03
CAJT9050-1CS 11201.84 CAD
CAJT9050-1 JT9050-1 CAJT9050-03
Benzyl alcohol ≥99.5%, BAKER ANALYZED®
Benzyl alcohol

Lot analysis on label.

Formula: C₆H₅CH₂OH
Boiling Pt: 205 °C (1013 hPa)
Melting Pt: –15.3 °C
Density: 1.05 g/cm³ (20 °C)
Flash Pt: 101 °C
Storage Temperature: Ambient
MDL Number: MFCD00004599
CAS Number: 100-51-6
EINECS: 202-859-9
Merck Index: 13,01126
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b0c85c958ac64f8b86c5836e49f2c4354b22886d&vid=633ad635461465914e782d5a22480c9ff8f81bf7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Assay (C₆H₅CH₂OH) (by GC, corrected for water) ≥99.5 %
Color (APHA) ≤10
Residue after Ignition ≤0.005 %
Solubility in H₂O Passes Test
Chlorinated Compounds (as Cl) Passes Test
Water (H₂O)(by Karl Fischer titrn) ≤0.05 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR