Orthoboric acid ≥99.5% (by acidimetry), granular, Ultrapure Bioreagent for molecular biology, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Synonyms: Boron trihydroxide, Boron hydroxide, Boric acid, Boracic acid

Management of Change (MOC) category = N

BAKR4035-01 BAKR4035-05
CAJT4035-1EA 231.38 CAD
CAJT4035-1 CAJT4035-5
Orthoboric acid ≥99.5% (by acidimetry), granular, Ultrapure Bioreagent for molecular biology, J.T.Baker®
ortho-Boric acid

For molecular biology buffers. Lot analysis on label.

Formula: B(OH)₃
MW: 61.83 g/mol
Boiling Pt: 185
Melting Pt: 160
Density: 1.44 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011337
CAS Number: 10043-35-3
EINECS: 233-139-2
REACH: 01-2119486683-25
Merck Index: 13,01326
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=098dd700154daaf6338ebb270902bb2bc704f27d&vid=6eec7d529090dd4c4b966ed8cd8d057153c5a47b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Assay (H₃BO₃) (by acidimetry) ≥99.5 %
RNase Activity Passes Test
DNase Activity (None Detected) Passes Test
Protease Activity Passes Test
Chloride (Cl) ≤0.001 %
Phosphate (PO₄) ≤0.001 %
Sulfate (SO₄) ≤0.005 %
Calcium (Ca) ≤0.005 %
Heavy Metals (as Pb) ≤0.001 %
Insoluble in Methanol ≤0.005 %
Trace Impurities - Iron (Fe) ≤0.001 %
Trace Impurities - Arsenic (As) ≤1.000 ppm
Trace Impurities - Magnesium (Mg) ≤5.000 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR