Diboron trioxide ≥98.0%, purified, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
|
Danger
|
Synonyms:
Boron oxide, Boric anhydride
Management of Change (MOC) category = N
BAKR1176-05
CAJT1176-5CS
4501.96
CAD
CAJT1176-5
Diboron trioxide ≥98.0%, purified, J.T.Baker®
di-Boron trioxide
Powder.
Formula:
B₂O₃ Boiling Pt: 1860 °C (1013 hPa) Melting Pt: 450 °C Density: 1.81…1.86 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011315 CAS Number: 1303-86-2 EINECS: 215-125-8 REACH: 01-2119486655-24 Merck Index: 13,01327 |
Specification Test Results
For Laboratory,Research,or Manufacturing Use | |
Assay (B₂O₃) (ignited basis) | ≥98.0 % |
Loss on Ignition | ≤10.0 % |
Sulfate (SO₄) | ≤0.05 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...