Bismuth(III) nitrate pentahydrate ≥98.0% (by EDTA titration), crystals, BAKER ANALYZED® ACS

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Synonyms: Bismuth trinitrate pentahydrate

Management of Change (MOC) category = N

BAKR1092-01 1092-04
CAJT1092-1CS 2780.23 CAD
CAJT1092-1 JT1092-4
Bismuth(III) nitrate pentahydrate ≥98.0% (by EDTA titration), crystals, BAKER ANALYZED® ACS
Bismuth (III) nitrate pentahydrate

Crystals. Lot analysis on label.

Formula: Bi(NO₃)₃·5H₂O
MW: 485.07 g/mol
Boiling Pt: 75…80 °C (1013 hPa)
Melting Pt: 30
Density: 2.83 g/cm³
MDL Number: MFCD00149157
CAS Number: 10035-06-0
EINECS: 233-791-8
UN: 1477
ADR: 5.1 II

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
ACS - Assay ((Bi(NO₃)₃·5H₂O)) (by EDTA titrn) ≥98.0 %
Insoluble in HNO₃ ≤0.005 %
Chloride (Cl) ≤0.001 %
Sulfate (SO₄) ≤0.005 %
Arsenic (As) ≤0.001 %
Calcium (Ca) ≤0.005 %
Copper (Cu) ≤0.002 %
Trace Impurities - Iron (Fe) ≤0.001 %
Lead (Pb) ≤0.002 %
Potassium (K) ≤0.01 %
Silver (Ag) ≤0.001 %
Sodium (Na) ≤0.02 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR