Barium carbonate ≥99% (by acidimetry), powder, BAKER ANANYZED™ ACS
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
|
Danger
|
Management of Change (MOC) category = N
BAKR0950-07
BAKR0950-R
0950-01
CAJT0950-7EA
4194.13
CAD
CAJT0950-7
CAJT0950-R
CAJT0950-5
JT0950-1
Barium carbonate ≥99% (by acidimetry), powder, BAKER ANANYZED™ ACS
Barium carbonate
Powder. Lot analysis on label.
Formula:
BaCO₃ Melting Pt: 1350 °C Density: 4.30 g/cm³ Storage Temperature: Ambient |
MDL Number:
MFCD00003448 CAS Number: 513-77-9 EINECS: 208-167-3 UN: 1564 ADR: 6.1,III Merck Index: 13,00972 |
Specification Test Results
Meets ACS Reagent Chemical Requirements | |
For Laboratory,Research,or Manufacturing Use | |
Assay (BaCO₃) (by acidimetry) | 99.0 - 101.0 % |
Insoluble in Dilute HCl | ≤0.015 % |
Chloride (Cl) | ≤0.002 % |
Water-Soluble Titrable Base, meq/g | ≤0.0020 |
Oxidizing Substances (as NO₃) | ≤0.005 % |
Sulfide (S) | ≤0.001 % |
Calcium (Ca) | ≤0.05 % |
Heavy Metals (as Pb) | ≤0.001 % |
Iron (Fe) | ≤0.002 % |
Potassium (K) | ≤0.005 % |
Sodium (Na) | ≤0.010 % |
Strontium (Sr) | ≤0.7 % |
Average Particle Diameter, µm (APD) | ≤5 |
Specific Surface Area, m²/g, For Information Only | |
Bulk Density (g/cc), For Information Only | |
Mesh (Wet Screen Analysis) - On U.S. No. 325 Sieve, For Information Only |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...