Orthoboric acid 99.5-100.5% (dry basis), granular NF, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC

Synonyms: Boric acidBoron hydroxideBoron trihydroxideBoracic acid

Management of Change (MOC) category = R

BAKR0091-05 BAKR0091-01 0091-01 0091-05
CAJT0091-1EA 545.12 CAD
JT0091-5 CAJT0091-1 CAJT0091-5 JT0091-1
Orthoboric acid 99.5-100.5% (dry basis), granular NF, J.T.Baker®
ortho-Boric acid
Formula: B(OH)₃
MW: 61.83 g/mol
Boiling Pt: 185 °C (1013 hPa)
Melting Pt: 160 °C
Density: 1.44 g/cm³ (25 °C)
MDL Number: MFCD00011337
CAS Number: 10043-35-3
Merck Index: 13,01326
https://ca.vwr.com/stibo/web/std.lang.all/05/54/16640554.jpg

Order Now

Specification Test Results

GMP Manufactured Product
Meets N.F. Requirements
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical
NF - Completeness of Solution Passes Test
NF - Identification Passes Test
NF - Solubility in Alcohol Passes Test
NF - Loss on Drying ≤0.5 %
NF - Assay (H₃BO₃) (dried basis) 99.5 - 100.5 %
Not for Internal Use
No Class 1,2,3 or other solvents are used or produced in the
manufacturing or purification of the product.
Elemental Impurities (USP 232, EP 5.20) - Information on elemental
impurities for this product is available on the associated Product
Regulatory Data Sheet and elemental impurity profile report.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR