1-Butanol ≥99.4% (by GC), GR ACS, Supelco®

Supplier: MilliporeSigma

Synonyms: n-ButanolPropyl carbinol1-Butyl alcoholButyl Alcohol1-Hydroxy butanen-Butyl Alcohol

BX1780-20 BX1780-5 BX1780-6 BX1780-5 BX1780-20 BX1780-6
CABX1780-6CS 3131.94 CAD
EM-BX1780-6 CABX1780-6 EM-BX1780-5 CABX1780-5 EM-BX1780-20 CABX1780-20
1-Butanol ≥99.4% (by GC), GR ACS, Supelco®
1-Butanol

Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs

Formula: H₃C(CH₂)₂CH₂OH
MW: 74.12 g/mol
Boiling Pt: 116…118 °C (1013 hPa)
Melting Pt: –89 °C
Density: 0.81 g/cm3 (20 °C)
Flash Pt: 30 °C
MDL Number: MFCD00002964
CAS Number: 71-36-3
UN: 1120
ADR: 3,III
Merck Index: 13,01539
https://ca.vwr.com/stibo/web/std.lang.all/07/44/730744.jpg

Order Now

Specification Test Results

Assay (GC) 99.4% min
Carbonyl compounds (as butyraldehyde) 0.01% max
Color (APHA) 10 max
Di-n-Butyl ether [(C4 H9)2 O] 0.2% max
Form Clear liquid
Residue after evaporation 0.005% max
Titrable acid 0.0008meq/g max
Water (H2O) 0.1% max

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR