1-Butanol, OmniSolv®, Supelco®
Supplier: MilliporeSigma
|
Danger
|
BX1777-1
BX1777-6
CABX1777-1CS
3567
CAD
CABX1777-1
CABX1777-6
1-Butanol, OmniSolv®, Supelco®
1-Butanol
For HPLC, spectrophotometry, and chromatography. UV cutoff 215nm. Lot analysis on label.
Formula:
H₃C(CH₂)₂CH₂OH Boiling Pt: 117.6 °C (1013 hPa) Melting Pt: –89 °C Density: 0.812 g/cm³ (20 °C) Flash Pt: 30 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002964 CAS Number: 71-36-3 EINECS: 200-751-6 UN: 1120 ADR: 3,III REACH: 01-2119484630-38 Merck Index: 13,01539 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...