Butyl acetate ≥99.5%, GR ACS, Supelco®

Supplier: MilliporeSigma
Danger

BX1735-1 BX1735-6
CABX1735-1CS 907 CAD
CABX1735-1 CABX1735-6
Butyl acetate ≥99.5%, GR ACS, Supelco®
Butyl acetate

Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs

Formula: CH₃COO(CH₂)₃CH₃
Boiling Pt: 126 °C (1013 hPa)
Melting Pt: –76 °C
Density: 0.875 g/cm³ (20 °C)
Flash Pt: 25 °C
Storage Temperature: Ambient
MDL Number: MFCD00009445
CAS Number: 123-86-4
EINECS: 204-658-1
UN: 1123
ADR: 3,III
Merck Index: 13,01534
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=403329de943f8cb2ea6cd9d73ee35957d2140aa7&vid=682ab82de177b2985e94089408f7efebde59926d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (GC) 99.5% min
Color (APHA) 10 max
Form Clear liquid
Infrared spectrum Conforms to standard
n-Butyl alcohol 0.2% max
Residue after evaporation 0.001% max
Substances darkened by sulfuric acid To pass test
Titrable acid 0.0016meq/g max
Water (H2O) 0.1% max


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR