1-Butanol ≥99.9%, B&J Brand™ for HPLC, for gas chromatography, for spectrophotometry, Burdick & Jackson™

Supplier: HONEYWELL BURDICK & JACKSON CA
Danger

Synonyms: n-ButanolPropyl carbinol1-Butyl alcoholButyl Alcohol1-Hydroxy butanen-Butyl Alcohol

024-4 024-1L 024-1L
CABJ024-1LCS 1987.77 CAD
BJ024-1L CABJ024-1L BJ024-4
1-Butanol ≥99.9%, B&J Brand™ for HPLC, for gas chromatography, for spectrophotometry, Burdick & Jackson™
1-Butanol

For HPLC, liquid and gas chromatography, pesticide residue analysis, spectophotometry, organic synthesis and combinatorial chemistry.


  • Minimal UV contaminants
  • High GC purity
  • Low water content
  • Outstanding lot-to-lot consistency

Formula: H₃C(CH₂)₂CH₂OH
MW: 74.12 g/mol
Boiling Pt: 117 °C
Melting Pt: –89 °C
Density: 0.81 g/cm³ (20 °C)
Flash Pt: 37 °C
Storage Temperature: Ambient
MDL Number: MFCD00002964
CAS Number: 71-36-3
UN: 1120
ADR: 3,III
Merck Index: 13,01539
https://ca.vwr.com/stibo/web/std.lang.all/07/44/730744.jpg

Order Now

Specification Test Results

GC Analysis 99 % min.
Refractive Index (20ºC) 1.3986 - 1.3994
UV Absorbance @ 225 nm 0.5 AU max.
UV Absorbance @ 250 nm 0.04 AU max.
UV Absorbance @ 275 nm 0.01 AU max.
UV Absorbance @ 300 nm 0.005 AU max.
UV Absorbance @ 400 nm 0.005 AU max.
UV Cutoff 204 - 215
Water by Karl Fischer Titration 0.03 % max.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR