Acetone ≥99.5%, ChromAR® for liquid chromatography, for UV spectrophotometry, Macron Fine Chemicals™
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
|
|
Synonyms:
2-Propanone
, DMK
, Propanone
, Dimethyl ketone
Management of Change (MOC) category = N
MACR2435-06
2435-06
MACR2435-10
MK243510CS
1636.75
CAD
MK243510
CAMK243506
MK243506
Acetone ≥99.5%, ChromAR® for liquid chromatography, for UV spectrophotometry, Macron Fine Chemicals™
Acetone
ChromAR™ solvents let you consolidate the number of solvent grades used in a diverse variety of applications, saving money and storage space without compromising quality and reliability of test results. They’re tested for key analytical parameters in HPLC and spectrophotometry.
Formula:
CH₃COCH₃ MW: 58.08 g/mol Boiling Pt: 56.2 °C (1013 hPa) Melting Pt: –95.4 °C Density: 0.792 g/cm³ (20 °C) Flash Pt: <–20 °C (closed cup) |
MDL Number:
MFCD00008765 CAS Number: 67-64-1 UN: 1090 ADR: 3,II Merck Index: 13,00067 |
Specification Test Results
For Laboratory,Research,or Manufacturing Use | |
ACS - Assay (CH₃COCH₃) (by GC, corrected for water) | ≥99.5 % |
ACS - Aldehydes (as HCHO) | ≤0.002 % |
ACS - Color (APHA) | ≤10 |
ACS - Isopropyl Alcohol (CH₃CHOHCH₃)(by GC) | ≤0.05 % |
ACS - Methanol (CH₃OH)(by GC) | ≤0.05 % |
ACS - Residue after Evaporation | ≤0.0005 % |
ACS - Solubility in H₂O | Passes Test |
ACS - Substances Reducing Permanganate | Passes Test |
ACS - Titrable Acid (meq/g) | ≤0.0003 |
ACS - Titrable Base (meq/g) | ≤0.0006 |
ACS - Water (H₂O) | ≤0.5 % |
Ultraviolet Absorbance (1.00-cm cell vs. water) - 330 nm | ≤1.00 |
Ultraviolet Absorbance (1.00-cm cell vs. water) - 340 nm | ≤0.10 |
Ultraviolet Absorbance (1.00-cm cell vs. water) - 350 nm | ≤0.01 |
Ultraviolet Absorbance (1.00-cm cell vs. water) - 375 nm | ≤0.01 |
Ultraviolet Absorbance (1.00-cm cell vs. water) - 400 nm | ≤0.010 |
Filtered through a 0.2 micron filter. | |
Appearance (clear, colorless liquid) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...