Acetone ≥99.5%, UltimAR® for HPLC, for gas chromatography, Macron Fine Chemicals™
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
|
Danger
|
Synonyms:
2-Propanone, DMK, Propanone, Dimethyl ketone
Management of Change (MOC) category = NI
MACRH451-06
MACRH451-10
CAMKH45106CS
1669.62
CAD
CAMKH45106
CAMKH45110
Acetone ≥99.5%, UltimAR® for HPLC, for gas chromatography, Macron Fine Chemicals™
Acetone
UltimAR—Solvents that meet ACS specifications for liquid chromatography, UV-spectrophotometry, and general purpose use. They are also tested by GC/ECD and GC/FID for use in extraction/concentration sample clean-up protocols for trace-level organic impurities.
Formula:
CH₃COCH₃ MW: 58.08 g/mol Boiling Pt: 56.2 Melting Pt: –95.4 °C Density: 0.792 g/cm³ (20 °C) Flash Pt: <–20 °C (closed cup) Storage Temperature: Ambient |
MDL Number:
MFCD00008765 CAS Number: 67-64-1 EINECS: 200-662-2 UN: 1090 ADR: 3 II REACH: 01-2119471330-49 Merck Index: 13,00067 |
|
Specification Test Results
For Laboratory,Research,or Manufacturing Use | |
ACS - Assay (CH₃COCH₃) (by GC, corrected for water) | ≥99.5 % |
ACS - Aldehydes (as HCHO) | ≤0.002 % |
ACS - Color (APHA) | ≤10 |
ACS - Isopropyl Alcohol (CH₃CHOHCH₃)(by GC) | ≤0.05 % |
ACS - Methanol (CH₃OH)(by GC) | ≤0.05 % |
ACS - Residue after Evaporation | ≤0.0001 % |
ACS - Solubility in H₂O | Passes Test |
ACS - Substances Reducing Permanganate | Passes Test |
ACS - Titrable Acid (meq/g) | ≤0.0003 |
ACS - Titrable Base (meq/g) | ≤0.0006 |
ACS - Water (H₂O) | ≤0.5 % |
Optical Absorbance (1-cm path vs water) - 330 nm | ≤1.00 |
Optical Absorbance (1-cm path vs water) - 340 nm | ≤0.06 |
Optical Absorbance (1-cm path vs water) - 400 - 350 nm | ≤0.01 |
Fluorescence Trace Impurities, measured as Quinine Base - at Emission Maximum for Impurities | ≤1.0 ppb |
ECD Sensitive Impurities (as Heptachl or Epoxide) Single Peak (ng/L) | ≤10 ppb |
FID Sensitive Impurities (as 2-Octano l) Single Impurity Peak (ng/mL) | ≤5 ppb |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...